Methyl 2-(3-(benzyloxy)phenyl)acetate ≥98%

Supplier: Ambeed
Warning

A1396724-1G A1396724-250MG A1396724-5G
77592-650EA 48.18 USD
77592-650 77597-134 77597-136
Methyl 2-(3-(benzyloxy)phenyl)acetate ≥98%
Methyl 2-(3-(benzyloxy)phenyl)acetate

Precaución: for Research Use Only

Formula: C₁₆H₁₆O₃
MW: 256.2964 g/mol
MDL Number: MFCD03093835
CAS Number: 62969-42-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR