Ethyl 2-(trans-4-aminocyclohexyl)acetate ≥95%

Supplier: Ambeed
Warning

Synonyms: Ethyltrans-2-(4-Aminocyclohexyl)acetate

A139608-100MG A139608-1G A139608-5G A139608-250MG
77600-022EA 43.05 USD
77600-022 77600-024 77600-026 77611-268
Ethyl 2-(trans-4-aminocyclohexyl)acetate ≥95%
Ethyl 2-(trans-4-aminocyclohexyl)acetate

Precaución: for Research Use Only

Formula: C₁₀H₁₉NO₂
MDL Number: MFCD01333995
CAS Number: 76308-28-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR