9-cis-Retinoic acid ≥97%

Supplier: Ambeed
Danger

Synonyms: 9-cis-RA, NSC-659772, Alitretinoin, Panretin, (2E,4E,6Z,8E)-3,7-Dimethyl-9-(2,6,6-trimethylcyclohex-1-en-1-yl)nona-2,4,6,8-tetraenoic acid

A1396006-10MG A1396006-1MG A1396006-5MG
77597-128EA 192.7 USD
77597-128 77597-130 77597-132
9-cis-Retinoic acid ≥97%
9-cis-Retinoic acid

Precaución: for Research Use Only

Formula: C₂₀H₂₈O₂
MW: 300.442 g/mol
MDL Number: MFCD00270072
CAS Number: 5300-03-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR