3,5-Difluoro-4-(hydroxymethyl)benzonitrile ≥97%
Supplier: Ambeed
|
Warning
|
A139350-1G
A139350-10G
A139350-250MG
A139350-25G
77667-012EA
49
USD
77667-012
77871-669
77871-670
77871-671
3,5-Difluoro-4-(hydroxymethyl)benzonitrile ≥97%
3,5-Difluoro-4-(hydroxymethyl)benzonitrile
Precaución: for Research Use Only
Formula:
C8H5F2NO |
MDL Number:
MFCD11847159 CAS Number: 228421-83-8 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...