2-(2-Hydroxyphenyl)-1H-benzimidazole ≥98%

Supplier: Ambeed
Warning

A139059-25G A139059-1G
77611-260EA 272.95 USD
77611-260 77885-174
2-(2-Hydroxyphenyl)-1H-benzimidazole ≥98%
2-(2-Hydroxyphenyl)-1H-benzimidazole

Precaución: for Research Use Only

Formula: C₁₃H₁₀N₂O
MDL Number: MFCD00022668
CAS Number: 2963-66-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR