Manganese citrate decahydrate ≥98%

Supplier: Ambeed
Warning

A139033-100G A139033-1G A139033-25G A139033-5G
77878-980EA 199 USD
77878-980 77878-981 77878-982 77878-983
Manganese citrate decahydrate ≥98%
Manganese citrate decahydrate

Precaución: for Research Use Only

Formula: C₁₂H₃₀Mn₃O₂₄
CAS Number: 2828433-42-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR