2-Methoxy-3-nitro-5-pyridineboronic acid pinacol ester ≥95%

Supplier: Ambeed
Warning

Synonyms: 2-Methoxy-3-nitropyridine-5-boronic acid pinacol ester, 2-Methoxy-3-nitro-5-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)pyridine

A138986-250MG A138986-100MG A138986-1G A138986-5G
77600-014EA 47.15 USD
77600-014 77667-000 77667-002 77667-004
2-Methoxy-3-nitro-5-pyridineboronic acid pinacol ester ≥95%
2-Methoxy-3-nitro-5-pyridineboronic acid pinacol ester

Precaución: for Research Use Only

Formula: C₁₂H₁₇BN₂O₅
MW: 280.08 g/mol
MDL Number: MFCD12923397
CAS Number: 1083168-94-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR