6-Chloro-5-methyl-1-(tetrahydro-2H-pyran-2-yl)-4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)-1H-indazole ≥98%

Supplier: Ambeed
Warning

A1388430-100MG A1388430-250MG A1388430-1G
77592-638EA 189.63 USD
77592-638 77592-640 77622-012
6-Chloro-5-methyl-1-(tetrahydro-2H-pyran-2-yl)-4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)-1H-indazole ≥98%
6-Chloro-5-methyl-1-(tetrahydro-2H-pyran-2-yl)-4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)-1H-indazole

Precaución: for Research Use Only

Formula: C₁₉H₂₆BClN₂O₃
MW: 376.69 g/mol
MDL Number: MFCD33398825
CAS Number: 2374152-83-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR