Ethyl 4-(trifluoromethyl)-1H-pyrazole-3-carboxylate ≥98%
Supplier: Ambeed
|
Warning
|
A138631-1G
A138631-100MG
A138631-250MG
77597-102EA
527.88
USD
77597-102
77615-104
77747-983
Ethyl 4-(trifluoromethyl)-1H-pyrazole-3-carboxylate ≥98%
Ethyl 4-(trifluoromethyl)-1H-pyrazole-3-carboxylate
Precaución: for Research Use Only
Formula:
C₇H₇F₃N₂O₂ MW: 208.1400 g/mol |
MDL Number:
MFCD20486284 CAS Number: 934758-94-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...