Chloro-IB-MECA ≥99%

Supplier: Ambeed
Warning

Synonyms: Cl-IB-MECA, 2-Chloro-N6-(3-iodobenzyl)adenosine-5'-N-methyluronamide, Namodenoson, (2S,3S,4R,5R)-5-(2-Chloro-6-((3-iodobenzyl)amino)-9H-purin-9-yl)-3,4-dihydroxy-N-methyltetrahydrofuran-2-carboxamide

A138589-5MG A138589-10MG A138589-1MG A138589-2MG
77615-102EA 33.99 USD
77615-102 77745-985 77745-986 77745-987
Chloro-IB-MECA ≥99%
Chloro-IB-MECA

Precaución: for Research Use Only

Formula: C₁₈H₁₈ClIN₆O₄
MW: 544.73 g/mol
MDL Number: MFCD01861178
CAS Number: 163042-96-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR