(4-(Methyl(phenyl)carbamoyl)phenyl)boronic acid ≥98%

Supplier: Ambeed
Warning

A1383557-100MG A1383557-250MG
77867-763EA 15 USD
77867-763 77867-764
(4-(Methyl(phenyl)carbamoyl)phenyl)boronic acid ≥98%
(4-(Methyl(phenyl)carbamoyl)phenyl)boronic acid

Precaución: for Research Use Only

Formula: C₁₄H₁₄BNO₃
MDL Number: MFCD32687510
CAS Number: 874219-49-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR