Methyl 2-hydroxy-6-nitrobenzoate ≥97%

Supplier: Ambeed
Warning

A138247-250MG A138247-100MG
77670-978EA 237 USD
77670-978 77883-114
Methyl 2-hydroxy-6-nitrobenzoate ≥97%
Methyl 2-hydroxy-6-nitrobenzoate

Precaución: for Research Use Only

Formula: C₈H₇NO₅
MW: 197.14 g/mol
MDL Number: MFCD00017565
CAS Number: 1261504-50-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR