Ethyl 6-(aminomethyl)nicotinate hydrochloride ≥95%

Supplier: Ambeed
Warning

A138035-1G A138035-250MG
77683-210EA 79 USD
77683-210 77683-211
Ethyl 6-(aminomethyl)nicotinate hydrochloride ≥95%
Ethyl 6-(aminomethyl)nicotinate hydrochloride

Precaución: For research use only.

Formula: C₉H₁₂N₂O₂·ClH
MW: 216.665 g/mol
MDL Number: MFCD09788628
CAS Number: 1189434-55-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR