(Z)-2-((Furan-2-ylmethyl)sulfinyl)-N-(4-((4-(piperidin-1-ylmethyl)pyridin-2-yl)oxy)but-2-en-1-yl)acetamide ≥97%

Supplier: Ambeed
Warning

Synonyms: (Z)-Lafutidine

A1380188-100MG A1380188-1G A1380188-250MG
77621-982EA 24.72 USD
77621-982 77621-984 77621-986
(Z)-2-((Furan-2-ylmethyl)sulfinyl)-N-(4-((4-(piperidin-1-ylmethyl)pyridin-2-yl)oxy)but-2-en-1-yl)acetamide ≥97%
(Z)-2-((Furan-2-ylmethyl)sulfinyl)-N-(4-((4-(piperidin-1-ylmethyl)pyridin-2-yl)oxy)but-2-en-1-yl)acetamide

Precaución: for Research Use Only

Formula: C₂₂H₂₉N₃O₄S
MW: 431.5484 g/mol
CAS Number: 206449-93-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR