Benzyl (2,3-difluoro-5-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)phenyl)carbamate ≥98%

Supplier: Ambeed
Warning

A1379579-100MG A1379579-1G A1379579-250MG
77867-745EA 33 USD
77867-745 77867-746 77867-747
Benzyl (2,3-difluoro-5-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)phenyl)carbamate ≥98%
Benzyl (2,3-difluoro-5-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)phenyl)carbamate

Precaución: for Research Use Only

Formula: C₂₀H₂₂BF₂NO₄
MW: 389.2 g/mol
MDL Number: MFCD32065769
CAS Number: 2377607-45-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR