4-Chloro-3-fluoropicolinic acid ≥98%

Supplier: Ambeed
Warning

A137953-100MG A137953-1G A137953-250MG
77889-931EA 11 USD
77889-931 77889-932 77889-933
4-Chloro-3-fluoropicolinic acid ≥98%
4-Chloro-3-fluoropicolinic acid

Precaución: for Research Use Only

Formula: C₆H₃ClFNO₂
MDL Number: MFCD16249576
CAS Number: 860296-21-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR