(3-(((Benzyloxy)carbonyl)amino)-4,5-difluorophenyl)boronic acid ≥98%

Supplier: Ambeed
Warning

A1379111-100MG A1379111-250MG
77867-743EA 32 USD
77867-743 77867-744
(3-(((Benzyloxy)carbonyl)amino)-4,5-difluorophenyl)boronic acid ≥98%
(3-(((Benzyloxy)carbonyl)amino)-4,5-difluorophenyl)boronic acid

Precaución: for Research Use Only

Formula: C₁₄H₁₂BF₂NO₄
MDL Number: MFCD32065670
CAS Number: 2377609-82-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR