(3AS,4S,6aR)-N-[3-(11,12-Didehydrodibenz[b,f]azocin-5(6H)-yl)-3-oxopropyl]hexahydro-2-oxo-1H-thieno[3,4-d]imidazole-4-pentanamide ≥98%

Supplier: Ambeed
Warning

A1378913-100MG A1378913-25MG A1378913-50MG A1378913-250MG
77597-094EA 428.45 USD
77597-094 77621-978 77621-980 77867-742
(3AS,4S,6aR)-N-[3-(11,12-Didehydrodibenz[b,f]azocin-5(6H)-yl)-3-oxopropyl]hexahydro-2-oxo-1H-thieno[3,4-d]imidazole-4-pentanamide ≥98%
(3AS,4S,6aR)-N-[3-(11,12-Didehydrodibenz[b,f]azocin-5(6H)-yl)-3-oxopropyl]hexahydro-2-oxo-1H-thieno[3,4-d]imidazole-4-pentanamide

Precaución: for Research Use Only

Formula: C₂₈H₃₀N₄O₃S
MDL Number: MFCD22380751
CAS Number: 1418217-95-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR