11,12-Didehydro-δ-oxodibenz[b,f]azocine-5(6H)-pentanoic acid ≥98%

Supplier: Ambeed
Warning

Synonyms: DBCO-C3-acid

A1378911-100MG A1378911-250MG
77592-624EA 127.1 USD
77592-624 77592-626
11,12-Didehydro-δ-oxodibenz[b,f]azocine-5(6H)-pentanoic acid ≥98%
11,12-Didehydro-δ-oxodibenz[b,f]azocine-5(6H)-pentanoic acid

Precaución: for Research Use Only

Formula: C₂₀H₁₇NO₃
MW: 319.35 g/mol
MDL Number: MFCD30748340
CAS Number: 1207355-31-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR