Ethyl 2-(2-(N-(2,4-dimethoxyphenyl)propiolamido)-2-(thiophen-2-yl)acetamido)acetate ≥98%

Supplier: Ambeed
Danger

Synonyms: PACMA 31

A1378788-10MG A1378788-1MG A1378788-5MG
77597-088EA 153.75 USD
77597-088 77597-090 77597-092
Ethyl 2-(2-(N-(2,4-dimethoxyphenyl)propiolamido)-2-(thiophen-2-yl)acetamido)acetate ≥98%
Ethyl 2-(2-(N-(2,4-dimethoxyphenyl)propiolamido)-2-(thiophen-2-yl)acetamido)acetate

Precaución: for Research Use Only

Formula: C₂₁H₂₂N₂O₆S
MW: 430.47 g/mol
MDL Number: MFCD26793891
CAS Number: 1401089-31-3
UN: 2811
ADR: 6.1,Ⅲ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR