(S)-2-((((9H-Fluoren-9-yl)methoxy)carbonyl)amino)-5-(tert-butoxy)-2-methyl-5-oxopentanoic acid ≥98%

Supplier: Ambeed
Warning

A1378245-100MG A1378245-250MG
77621-974EA 87.55 USD
77621-974 77621-976
(S)-2-((((9H-Fluoren-9-yl)methoxy)carbonyl)amino)-5-(tert-butoxy)-2-methyl-5-oxopentanoic acid ≥98%
(S)-2-((((9H-Fluoren-9-yl)methoxy)carbonyl)amino)-5-(tert-butoxy)-2-methyl-5-oxopentanoic acid

Precaución: for Research Use Only

Formula: C₂₅H₂₉NO₆
MW: 439.5 g/mol
MDL Number: MFCD32899204
CAS Number: 1072845-48-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR