tert-Butyl [3,3'-biazetidine]-1-carboxylate hemioxalate ≥98%

Supplier: Ambeed
Warning

Synonyms: oxalicacidbis(tert-butyl3-(azetidin-3-yl)azetidine-1-carboxylate)

A1377707-100MG A1377707-250MG A1377707-50MG
77621-964EA 241.02 USD
77621-964 77621-966 77621-968
tert-Butyl [3,3'-biazetidine]-1-carboxylate hemioxalate ≥98%
tert-Butyl [3,3'-biazetidine]-1-carboxylate hemioxalate

Precaución: for Research Use Only

Formula: C₂₄H₄₂N₄O₈
MW: 514.61 g/mol
MDL Number: MFCD30497623
CAS Number: 2059937-79-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR