(4-Hydroxyphenyl)(methyl)(naphthalen-1-ylmethyl)sulfonium hexafluorostibate(V) ≥98%

Supplier: Ambeed
Danger

Synonyms: Sanaid SI 60

A1377211-1G A1377211-250MG A1377211-100MG
77597-076EA 237.8 USD
77597-076 77597-078 77621-962
(4-Hydroxyphenyl)(methyl)(naphthalen-1-ylmethyl)sulfonium hexafluorostibate(V) ≥98%
(4-Hydroxyphenyl)(methyl)(naphthalen-1-ylmethyl)sulfonium hexafluorostibate(V)

Precaución: for Research Use Only

Formula: C₁₈H₁₇F₆OSSb
MW: 517.14 g/mol
CAS Number: 133152-67-7
UN: 3077
ADR: 9 Ⅲ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR