3-((1H-Imidazol-5-yl)methylene)-5-methoxyindolin-2-one 99+%

Supplier: Ambeed
Warning

Synonyms: SU-9516

A137702-1MG A137702-5MG
77745-979EA 22 USD
77745-979 77745-980
3-((1H-Imidazol-5-yl)methylene)-5-methoxyindolin-2-one 99+%
3-((1H-Imidazol-5-yl)methylene)-5-methoxyindolin-2-one

Precaución: for Research Use Only

Formula: C₁₃H₁₁N₃O₂
MW: 241.25 g/mol
MDL Number: MFCD17010284
CAS Number: 666837-93-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR