CPI-0610 ≥98%

Supplier: Ambeed
Warning

Synonyms: Pelabresib, (S)-2-(6-(4-chlorophenyl)-1-methyl-4H-benzo[c]isoxazolo[4,5-e]azepin-4-yl)acetamide

A137689-10MG A137689-5MG
77745-977EA 150 USD
77745-977 77745-978
CPI-0610 ≥98%
CPI-0610

Precaución: for Research Use Only

Formula: C₂₀H₁₆ClN₃O₂
MW: 365.82 g/mol
MDL Number: MFCD28144505
CAS Number: 1380087-89-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR