N-(3-Hydroxy-2-(4-(4-methyl-1,4-diazepan-1-yl)benzamido)phenyl)-4-methoxybenzamide ≥97%
Supplier: Ambeed
|
Warning
|
Synonyms:
Darexaban
A1376680-10MG
A1376680-1MG
A1376680-5MG
77597-060EA
92.25
USD
77597-060
77597-062
77597-064
N-(3-Hydroxy-2-(4-(4-methyl-1,4-diazepan-1-yl)benzamido)phenyl)-4-methoxybenzamide ≥97%
N-(3-Hydroxy-2-(4-(4-methyl-1,4-diazepan-1-yl)benzamido)phenyl)-4-methoxybenzamide
Precaución: for Research Use Only
Formula:
C₂₇H₃₀N₄O₄ MW: 474.55 g/mol |
MDL Number:
MFCD16620506 CAS Number: 365462-23-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...