1,1,3,3-Tetramethyl-2-thiourea 99+%

Supplier: Ambeed
Warning

A1375375-25G A1375375-500G
77867-716EA 7 USD
77867-716 77867-717
1,1,3,3-Tetramethyl-2-thiourea 99+%
1,1,3,3-Tetramethyl-2-thiourea

Precaución: for Research Use Only

Formula: C₅H₁₂N₂S
MDL Number: MFCD00008324
CAS Number: 2782-91-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR