5-Fluoro-1H-indole-2-carbaldehyde ≥98%

Supplier: Ambeed
Warning

A137381-1G A137381-100MG
77611-246EA 195.14 USD
77611-246 77889-924
5-Fluoro-1H-indole-2-carbaldehyde ≥98%
5-Fluoro-1H-indole-2-carbaldehyde

Precaución: for Research Use Only

Formula: C₉H₆FNO
MDL Number: MFCD06800739
CAS Number: 220943-23-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR