1-(4-(Benzo[d]thiazol-2-ylmethoxy)-2-methylphenyl)-3-(3,4-dichlorophenyl)urea ≥97%

Supplier: Ambeed
Warning

A1373802-100MG A1373802-50MG
77621-930EA 139.05 USD
77621-930 77621-932
1-(4-(Benzo[d]thiazol-2-ylmethoxy)-2-methylphenyl)-3-(3,4-dichlorophenyl)urea ≥97%
1-(4-(Benzo[d]thiazol-2-ylmethoxy)-2-methylphenyl)-3-(3,4-dichlorophenyl)urea

Precaución: for Research Use Only

Formula: C₂₂H₁₇Cl₂N₃O₂S
MDL Number: MFCD04092440
CAS Number: 724453-98-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR