2-Chloro-3-methyl-3H-pyrrolo[3,2-d]pyrimidin-4(5H)-one ≥97%

Supplier: Ambeed
Warning

A1373628-250MG A1373628-100MG A1373628-1G
77592-604EA 76.88 USD
77592-604 77597-040 77597-042
2-Chloro-3-methyl-3H-pyrrolo[3,2-d]pyrimidin-4(5H)-one ≥97%
2-Chloro-3-methyl-3H-pyrrolo[3,2-d]pyrimidin-4(5H)-one

Precaución: for Research Use Only

Formula: C₇H₆ClN₃O
MW: 183.5950 g/mol
MDL Number: MFCD31924860
CAS Number: 2351929-82-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR