Perylen-3-ylboronic acid ≥98%
Supplier: Ambeed
|
Warning
|
A137358-250MG
A137358-1G
77615-088EA
121.54
USD
77615-088
77747-972
Perylen-3-ylboronic acid ≥98%
Perylen-3-ylboronic acid
Precaución: for Research Use Only
Formula:
C₂₀H₁₃BO₂ MW: 296.1270 g/mol |
MDL Number:
MFCD12031332 CAS Number: 955121-20-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...