trans-2-(Boc-amino)cyclopropanemethanol ≥98%

Supplier: Ambeed
Warning

Synonyms: rel-tert-Butyl ((1R,2R)-2-(hydroxymethyl)cyclopropyl)carbamate

A1373498-1G A1373498-250MG
77867-692EA 982 USD
77867-692 77867-693
trans-2-(Boc-amino)cyclopropanemethanol ≥98%
trans-2-(Boc-amino)cyclopropanemethanol

Precaución: for Research Use Only

Formula: C₉H₁₇NO₃
MDL Number: MFCD03844612
CAS Number: 170299-53-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR