[2,2':6',2''-Terpyridin]-4'-ylmethanol ≥98%

Supplier: Ambeed
Warning

A1372943-100MG A1372943-1G A1372943-250MG A1372943-5G
77597-032EA 13.33 USD
77597-032 77597-034 77597-036 77867-686
[2,2':6',2''-Terpyridin]-4'-ylmethanol ≥98%
[2,2':6',2''-Terpyridin]-4'-ylmethanol

Precaución: for Research Use Only

Formula: C₁₆H₁₃N₃O
MDL Number: MFCD14636293
CAS Number: 148332-32-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR