4-Methylvaleraldehyde ≥98%

Supplier: Ambeed
Danger

Synonyms: Isocaproaldehyde

A137237-250MG A137237-1G A137237-100MG
77600-000EA 237.8 USD
77600-000 77666-974 77666-972
4-Methylvaleraldehyde ≥98%
4-Methylvaleraldehyde

Precaución: for Research Use Only

Formula: C₆H₁₂O
MW: 100.16 g/mol
MDL Number: MFCD00871507
CAS Number: 1119-16-0
UN: 1989
ADR: 3,Ⅱ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR