ASK1-IN-1 ≥98%

Supplier: Ambeed
Warning

A1372186-25MG A1372186-100MG A1372186-50MG
77621-918EA 160.68 USD
77621-918 77867-676 77867-677
ASK1-IN-1 ≥98%
ASK1-IN-1

Precaución: for Research Use Only

Formula: C₁₉H₁₉N₉O₂
CAS Number: 2411382-24-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR