Ni(4-CF3stb)3 ≥98%

Supplier: Ambeed
Danger

A1372162-1G A1372162-250MG A1372162-5G
77597-022EA 128.13 USD
77597-022 77597-024 77621-916
Ni(4-CF3stb)3 ≥98%
Ni(4-CF3stb)3

Precaución: for Research Use Only

Formula: C₄₈H₃₀F₁₈Ni
MW: 1007.4165 g/mol
MDL Number: MFCD33404403
CAS Number: 2413906-36-0
UN: 2811
ADR: 6.1,Ⅲ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR