4-Aminobenzenesulfonamide hydrochloride ≥98%

Supplier: Ambeed
Warning

A1371181-1G A1371181-5G
77867-654EA 17 USD
77867-654 77867-655
4-Aminobenzenesulfonamide hydrochloride ≥98%
4-Aminobenzenesulfonamide hydrochloride

Precaución: for Research Use Only

Formula: C₆H₉ClN₂O₂S
MDL Number: MFCD01683072
CAS Number: 6101-31-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR