(2S,3R,4S)-3-Ethenyl-2-(β-D-glucopyranosyloxy)-3,4-dihydro-4-[2-[(3,4,5-trihydroxybenzoyl)oxy]ethyl]-2H-pyran-5-carboxylic acid methyl ester ≥98%

Supplier: Ambeed
Warning

A1371131-10MG A1371131-1MG A1371131-5MG
77597-012EA 74.82 USD
77597-012 77597-014 77597-016
(2S,3R,4S)-3-Ethenyl-2-(β-D-glucopyranosyloxy)-3,4-dihydro-4-[2-[(3,4,5-trihydroxybenzoyl)oxy]ethyl]-2H-pyran-5-carboxylic acid methyl ester ≥98%
(2S,3R,4S)-3-Ethenyl-2-(β-D-glucopyranosyloxy)-3,4-dihydro-4-[2-[(3,4,5-trihydroxybenzoyl)oxy]ethyl]-2H-pyran-5-carboxylic acid methyl ester

Precaución: for Research Use Only

Formula: C₂₄H₃₀O₁₄
MDL Number: MFCD21606970
CAS Number: 131189-57-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR