N-(Pyridazin-3-yl)-4-(3-((5-(trifluoromethyl)pyridin-2-yl)oxy)benzylidene)piperidine-1-carboxamide ≥98%

Supplier: Ambeed
Warning

Synonyms: Redafamdastat

A137082-10MG A137082-5MG
77685-182EA 42 USD
77685-182 77685-183
N-(Pyridazin-3-yl)-4-(3-((5-(trifluoromethyl)pyridin-2-yl)oxy)benzylidene)piperidine-1-carboxamide ≥98%
N-(Pyridazin-3-yl)-4-(3-((5-(trifluoromethyl)pyridin-2-yl)oxy)benzylidene)piperidine-1-carboxamide

Precaución: for Research Use Only

Formula: C₂₃H₂₀F₃N₅O₂
MW: 455.43 g/mol
MDL Number: MFCD18782721
CAS Number: 1020315-31-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR