4-((14-Amino-3,6,9,12-tetraoxatetradecyl)amino)-2-(2,6-dioxopiperidin-3-yl)isoindoline-1,3-dione hydrochloride ≥95%

Supplier: Ambeed
Warning

Synonyms: Pomalidomide-PEG4-C2-NH2 HCl

A1369837-100MG A1369837-50MG
77621-862EA 153.47 USD
77621-862 77867-636
4-((14-Amino-3,6,9,12-tetraoxatetradecyl)amino)-2-(2,6-dioxopiperidin-3-yl)isoindoline-1,3-dione hydrochloride ≥95%
4-((14-Amino-3,6,9,12-tetraoxatetradecyl)amino)-2-(2,6-dioxopiperidin-3-yl)isoindoline-1,3-dione hydrochloride

Precaución: for Research Use Only

Formula: C₂₃H₃₃ClN₄O₈
MW: 528.98 g/mol
MDL Number: MFCD32705122
CAS Number: 2357105-92-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR