Potassium (2,2-difluorocyclopropyl)trifluoroborate ≥98%

Supplier: Ambeed
Warning

A1366637-250MG A1366637-100MG
77596-982EA 147.6 USD
77596-982 77621-848
Potassium (2,2-difluorocyclopropyl)trifluoroborate ≥98%
Potassium (2,2-difluorocyclopropyl)trifluoroborate

Precaución: for Research Use Only

Formula: C₃H₃BF₅K
MW: 183.96 g/mol
MDL Number: MFCD31615914
CAS Number: 2416056-30-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR