(2R,3R)-2-(3,4-Dihydroxyphenyl)-3,5-dihydroxy-7-(((2S,3R,4S,5S,6R)-3,4,5-trihydroxy-6-(hydroxymethyl)tetrahydro-2H-pyran-2-yl)oxy)chroman-4-one ≥95%

Supplier: Ambeed
Warning

Synonyms: Taxifolin 7-O-β-D-glucoside

A1365743-10MG A1365743-1MG A1365743-5MG A1365743-25MG A1365743-50MG
77621-842EA 293.55 USD
77621-842 77621-844 77621-846 77867-612 77867-613
(2R,3R)-2-(3,4-Dihydroxyphenyl)-3,5-dihydroxy-7-(((2S,3R,4S,5S,6R)-3,4,5-trihydroxy-6-(hydroxymethyl)tetrahydro-2H-pyran-2-yl)oxy)chroman-4-one ≥95%
(2R,3R)-2-(3,4-Dihydroxyphenyl)-3,5-dihydroxy-7-(((2S,3R,4S,5S,6R)-3,4,5-trihydroxy-6-(hydroxymethyl)tetrahydro-2H-pyran-2-yl)oxy)chroman-4-one

Precaución: for Research Use Only

Formula: C₂₁H₂₂O₁₂
MDL Number: MFCD34470777
CAS Number: 14292-40-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR