(R)-2-Oxo-N-(prop-2-yn-1-yl)thiazolidine-4-carboxamide ≥98%

Supplier: Ambeed
Warning

Synonyms: Cystathionine-γ-lyase-IN-1

A1365574-10MG A1365574-1MG A1365574-5MG
77596-966EA 55.35 USD
77596-966 77596-968 77596-970
(R)-2-Oxo-N-(prop-2-yn-1-yl)thiazolidine-4-carboxamide ≥98%
(R)-2-Oxo-N-(prop-2-yn-1-yl)thiazolidine-4-carboxamide

Precaución: for Research Use Only

Formula: C₇H₈N₂O₂S
MW: 184.22 g/mol
MDL Number: MFCD30729654
CAS Number: 2165706-30-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR