4-((S)-2-((S)-2-(6-(2,5-Dioxo-2,5-dihydro-1H-pyrrol-1-yl)hexanamido)-3-methylbutanamido)propanamido)benzyl (4-nitrophenyl) carbonate ≥98%

Supplier: Ambeed
Warning

Synonyms: MC-Val-Ala-PAB-PNP

A1365551-50MG A1365551-100MG A1365551-1G
77621-830EA 99.91 USD
77621-830 77867-602 77867-603
4-((S)-2-((S)-2-(6-(2,5-Dioxo-2,5-dihydro-1H-pyrrol-1-yl)hexanamido)-3-methylbutanamido)propanamido)benzyl (4-nitrophenyl) carbonate ≥98%
4-((S)-2-((S)-2-(6-(2,5-Dioxo-2,5-dihydro-1H-pyrrol-1-yl)hexanamido)-3-methylbutanamido)propanamido)benzyl (4-nitrophenyl) carbonate

Precaución: for Research Use Only

Formula: C₃₂H₃₇N₅O₁₀
MW: 651.6637 g/mol
MDL Number: MFCD32220341
CAS Number: 1639939-40-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR