4-(3,5-Dimethoxyphenyl)-N-(7-fluoro-3-methoxyquinoxalin-2-yl)piperazine-1-carboxamide ≥98%
Supplier: Ambeed
|
Warning
|
Synonyms:
Supinoxin
A1365338-10MG
A1365338-1MG
A1365338-5MG
77621-822EA
64.89
USD
77621-822
77621-824
77621-826
4-(3,5-Dimethoxyphenyl)-N-(7-fluoro-3-methoxyquinoxalin-2-yl)piperazine-1-carboxamide ≥98%
4-(3,5-Dimethoxyphenyl)-N-(7-fluoro-3-methoxyquinoxalin-2-yl)piperazine-1-carboxamide
Precaución: for Research Use Only
Formula:
C₂₂H₂₄FN₅O₄ MW: 441.46 g/mol |
MDL Number:
MFCD32004516 CAS Number: 888478-45-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...