4-(3,5-Dimethoxy-4-(piperazin-1-ylmethyl)phenyl)-2-methyl-2,7-naphthyridin-1(2H)-one dihydrochloride ≥98%

Supplier: Ambeed
Warning

A1365245-10MG A1365245-1MG A1365245-5MG
77596-950EA 165.02 USD
77596-950 77596-952 77596-954
4-(3,5-Dimethoxy-4-(piperazin-1-ylmethyl)phenyl)-2-methyl-2,7-naphthyridin-1(2H)-one dihydrochloride ≥98%
4-(3,5-Dimethoxy-4-(piperazin-1-ylmethyl)phenyl)-2-methyl-2,7-naphthyridin-1(2H)-one dihydrochloride

Precaución: for Research Use Only

Formula: C₂₂H₂₈Cl₂N₄O₃
MDL Number: MFCD32062734
CAS Number: 2448414-48-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR