(S)-Pro-xylane ≥98%

Supplier: Ambeed
Warning

A1365228-1G A1365228-25G
77596-944EA 11.28 USD
77596-944 77867-590
(S)-Pro-xylane ≥98%
(S)-Pro-xylane

Precaución: for Research Use Only

Formula: C₈H₁₆O₅
MDL Number: MFCD32701904
CAS Number: 868156-46-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR