(S)-2-(2-(2-(6-(2,5-Dioxo-2,5-dihydro-1H-pyrrol-1-yl)hexanamido)acetamido)acetamido)-3-phenylpropanoic acid ≥98%

Supplier: Ambeed
Warning

Synonyms: MC-Gly-Gly-Phe

A1365096-10G A1365096-1G A1365096-250MG
77621-814EA 448.05 USD
77621-814 77621-816 77621-818
(S)-2-(2-(2-(6-(2,5-Dioxo-2,5-dihydro-1H-pyrrol-1-yl)hexanamido)acetamido)acetamido)-3-phenylpropanoic acid ≥98%
(S)-2-(2-(2-(6-(2,5-Dioxo-2,5-dihydro-1H-pyrrol-1-yl)hexanamido)acetamido)acetamido)-3-phenylpropanoic acid

Precaución: for Research Use Only

Formula: C₂₃H₂₈N₄O₇
MW: 472.49 g/mol
MDL Number: MFCD32690114
CAS Number: 1599440-15-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR