2-(Phenylamino)-N-((tetrahydro-2H-pyran-4-yl)methyl)-4-(trifluoromethyl)pyrimidine-5-carboxamide ≥98%

Supplier: Ambeed

Synonyms: CB2 modulator 1

A1364978-10MG A1364978-1MG A1364978-5MG
77596-936EA 94.3 USD
77596-936 77596-938 77596-940
2-(Phenylamino)-N-((tetrahydro-2H-pyran-4-yl)methyl)-4-(trifluoromethyl)pyrimidine-5-carboxamide ≥98%
2-(Phenylamino)-N-((tetrahydro-2H-pyran-4-yl)methyl)-4-(trifluoromethyl)pyrimidine-5-carboxamide

Precaución: for Research Use Only

Formula: C₁₈H₁₉F₃N₄O₂
MDL Number: MFCD32671340
CAS Number: 666261-80-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR