4-Bromo-6-((3,4-dichlorophenyl)thio)-1-(4-(dimethylcarbamoyl)benzyl)-1H-indole-2-carboxylic acid ≥98%

Supplier: Ambeed

A1364808-10MG A1364808-1MG A1364808-5MG
77596-930EA 190.65 USD
77596-930 77596-932 77596-934
4-Bromo-6-((3,4-dichlorophenyl)thio)-1-(4-(dimethylcarbamoyl)benzyl)-1H-indole-2-carboxylic acid ≥98%
4-Bromo-6-((3,4-dichlorophenyl)thio)-1-(4-(dimethylcarbamoyl)benzyl)-1H-indole-2-carboxylic acid

Precaución: for Research Use Only

Formula: C₂₅H₁₉BrCl₂N₂O₃S
MDL Number: MFCD31714165
CAS Number: 2216763-38-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR